3-(Chloromethyl)-5-thiophen-2-ylisoxazole 95+%
Supplier: Apollo Scientific
OR346418-1G
OR346418-500MG
OR346418-5G
APOSOR346418-1GEA
265
EUR
APOSOR346418-1G
APOSOR346418-500MG
APOSOR346418-5G
3-(Chloromethyl)-5-thiophen-2-ylisoxazole 95+%
3-(Chloromethyl)-5-thiophen-2-ylisoxazole
Formula:
C₈H₆ClNOS MW: 199,657 g/mol Melting Pt: 32…34 °C Storage Temperature: Ambient |
MDL Number:
MFCD07339176 CAS Number: 863669-57-2 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...