4-(Chloromethyl)-2-isopropylthiazole hydrochloride 95+%
Supplier: Apollo Scientific
OR346425-1G
OR346425-500MG
APOSOR346425-1GEA
317
EUR
APOSOR346425-1G
APOSOR346425-500MG
4-(Chloromethyl)-2-isopropylthiazole hydrochloride 95+%
4-(Chloromethyl)-2-isopropylthiazole hydrochloride
Formula:
C₇H₁₀ClNS·ClH MW: 212,14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07369243 CAS Number: 65386-28-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...