2-Vinylpyridine-3-carboxaldehyde 95+%

Supplier: Apollo Scientific

OR346434-500MG OR346434-5G
APOSOR346434-500MGEA 0 EUR
APOSOR346434-500MG APOSOR346434-5G
2-Vinylpyridine-3-carboxaldehyde 95+%
2-Vinylpyridine-3-carboxaldehyde
Formula: C₈H₇NO
MW: 133.147 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06654833
CAS Number: 893620-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR