2-(1-Ethoxy-propylidene)malononitrile 95+%

Supplier: Apollo Scientific

OR346445-1G OR346445-5G
APOSOR346445-1GEA 201 EUR
APOSOR346445-1G APOSOR346445-5G
2-(1-Ethoxy-propylidene)malononitrile 95+%
2-(1-Ethoxy-propylidene)malononitrile
Formula: C₈H₁₀N₂O
MW: 150,178 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09909756
CAS Number: 35260-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR