2-((E)-3-Ethoxy-2-methyl-allylidene)-malononitrile 95+%
Supplier: Apollo Scientific
OR346447-1G
OR346447-5G
APOSOR346447-1GEA
201
EUR
APOSOR346447-1G
APOSOR346447-5G
2-((E)-3-Ethoxy-2-methyl-allylidene)-malononitrile 95+%
2-((E)-3-Ethoxy-2-methyl-allylidene)-malononitrile
Formula:
C₉H₁₀N₂O MW: 162,188 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06214167 CAS Number: 65995-93-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...