3-Isopropylisoxazole-5-carboxaldehyde 95+%
Supplier: Apollo Scientific
OR346502-1G
OR346502-5G
APOSOR346502-1GEA
306
EUR
APOSOR346502-1G
APOSOR346502-5G
3-Isopropylisoxazole-5-carboxaldehyde 95+%
3-Isopropylisoxazole-5-carboxaldehyde
Formula:
C₇H₉NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD14702865 CAS Number: 121604-54-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...