3-(3-Azetidinyl)pyridine dihydrochloride 95+%

Supplier: Apollo Scientific

Synonyms: 3-(Azetidin-3-yl)pyridine dihydrochloride

OR346503-1G OR346503-250MG
APOSOR346503-1GEA 340 EUR
APOSOR346503-1G APOSOR346503-250MG
3-(3-Azetidinyl)pyridine dihydrochloride 95+%
3-(3-Azetidinyl)pyridine dihydrochloride
Formula: C₈H₁₀N₂·2ClH
MW: 207,1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20527356
CAS Number: 1236791-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR