4-Phenyloxazole 95+%
Supplier: Apollo Scientific
OR346522-1G
OR346522-5G
APOSOR346522-1GEA
183
EUR
APOSOR346522-1G
APOSOR346522-5G
4-Phenyloxazole 95+%
4-Phenyloxazole
Formula:
C₉H₇NO MW: 145,158 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11845045 CAS Number: 20662-89-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...