1-(4-Aminophenyl)-cyclopropanecarboxylic acid methyl ester 95+%

Supplier: Apollo Scientific

Synonyms: Methyl 1-(4-aMinophenyl)cyclopropanecarboxylate

OR346590-1G OR346590-250MG
APOSOR346590-1GEA 621 EUR
APOSOR346590-1G APOSOR346590-250MG
1-(4-Aminophenyl)-cyclopropanecarboxylic acid methyl ester 95+%
1-(4-Aminophenyl)-cyclopropanecarboxylic acid methyl ester
Formula: C₁₁H₁₃NO₂
MW: 191,226 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20693763
CAS Number: 824937-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR