(3-Methoxy-phenoxy)acetonitrile 95+%
Supplier: Apollo Scientific
OR346665-1G
OR346665-5G
APOSOR346665-1GEA
132
EUR
APOSOR346665-1G
APOSOR346665-5G
(3-Methoxy-phenoxy)acetonitrile 95+%
(3-Methoxy-phenoxy)acetonitrile
Formula:
C₉H₉NO₂ MW: 163,173 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01861402 CAS Number: 50635-23-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...