3-Chloropyridine-2-carboxamide 95+%

Supplier: Apollo Scientific

Synonyms: 3-Chloropicolinamide

OR346700-10G
APOSOR346700-10GEA 0 EUR
APOSOR346700-10G
3-Chloropyridine-2-carboxamide 95+%
3-Chloropyridine-2-carboxamide
Formula: C₆H₅ClN₂O
MW: 156,57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10574691
CAS Number: 114080-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR