4-(Pyridin-4-yl)benzoic acid methyl ester 95+%

Supplier: Apollo Scientific

Synonyms: Methyl 4-(pyridin-4-yl)benzoate

OR346705-1G OR346705-5G
APOSOR346705-1GEA 98.9 EUR
APOSOR346705-1G APOSOR346705-5G
4-(Pyridin-4-yl)benzoic acid methyl ester 95+%
4-(Pyridin-4-yl)benzoic acid methyl ester
Formula: C₁₃H₁₁NO₂
MW: 213,232 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04114575
CAS Number: 106047-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR