4-o-Tolylpyridine 90+%
Supplier: Apollo Scientific
OR346714-1G
OR346714-500MG
APOSOR346714-1GEA
622
EUR
APOSOR346714-1G
APOSOR346714-500MG
4-o-Tolylpyridine 90+%
4-o-Tolylpyridine
Formula:
C₁₂H₁₁N MW: 169,222 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00657927 CAS Number: 30456-66-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...