Ethyl 2-(4-chloro-2-(methylthio)pyrimidin-5-yl)acetate 97%

Supplier: Apollo Scientific

OR350175-1G OR350175-250MG
APOSOR350175-1GEA 107 EUR
APOSOR350175-1G APOSOR350175-250MG
Ethyl 2-(4-chloro-2-(methylthio)pyrimidin-5-yl)acetate 97%
Ethyl 2-(4-chloro-2-(methylthio)pyrimidin-5-yl)acetate
Formula: C₉H₁₁ClN₂O₂S
MW: 246,714 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11977460
CAS Number: 61727-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR