3-(5-Methyl-1,3,4-oxadiazol-2-yl)benzeneboronic acid 97%

Supplier: Apollo Scientific
Warning

Synonyms: 2-(3-Boronophenyl)-5-methyl-1,3,4-oxadiazole, (3-(5-Methyl-1,3,4-oxadiazol-2-yl)phenyl)boronic acid

OR3585-1G OR3585-250MG OR3585-50MG
APOSOR3585-1GEA 249 EUR
APOSOR3585-1G APOSOR3585-250MG APOSOR3585-50MG
3-(5-Methyl-1,3,4-oxadiazol-2-yl)benzeneboronic acid 97%
3-(5-Methyl-1,3,4-oxadiazol-2-yl)benzeneboronic acid
Formula: C₉H₉BN₂O₃
MW: 203,99 g/mol
Melting Pt: 160…162 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08689540
CAS Number: 913836-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR