4-(2-Pyridinyl)phenylboronic acid, pinacol ester 98%

Supplier: Apollo Scientific
Warning

Synonyms: 2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)pyridine

OR360137-10G OR360137-1G OR360137-5G
APOSOR360137-10GEA 285 EUR
APOSOR360137-10G APOSOR360137-1G APOSOR360137-5G
4-(2-Pyridinyl)phenylboronic acid, pinacol ester 98%
4-(2-Pyridinyl)phenylboronic acid, pinacol ester
Formula: C₁₇H₂₀BNO₂
MW: 281,157 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11973624
CAS Number: 908350-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR