(6-Aminopyridin-3-yl)boronic acid hydrochloride 95+%

Supplier: Apollo Scientific
Warning

Synonyms: 6-Aminopyridine-3-boronic acid hydrochloride

OR360171-5G OR360171-1G
APOSOR360171-5GEA 119 EUR
APOSOR360171-5G APOSOR360171-1G
(6-Aminopyridin-3-yl)boronic acid hydrochloride 95+%
(6-Aminopyridin-3-yl)boronic acid hydrochloride
Formula: C₅H₈BClN₂O₂
MW: 174,39 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12198146
CAS Number: 851524-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR