[4-[(Dimethylamino)methyl]phenyl]boronic acid 97%

Supplier: Apollo Scientific

Synonyms: 4-((Dimethylamino)methyl)phenylboronic acid

OR360262-5G OR360262-1G OR360262-250MG
APOSOR360262-5GEA 261 EUR
APOSOR360262-5G APOSOR360262-1G APOSOR360262-250MG
[4-[(Dimethylamino)methyl]phenyl]boronic acid 97%
[4-[(Dimethylamino)methyl]phenyl]boronic acid
Formula: C₉H₁₄BNO₂
MW: 179.024 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01319001
CAS Number: 70799-12-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR