3-Amino-5-cyanophenylboronic acid 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(3-Amino-5-cyanophenyl)boronic acid
OR360638-100MG
OR360638-1G
OR360638-250MG
APOSOR360638-100MGEA
74.1
EUR
APOSOR360638-100MG
APOSOR360638-1G
APOSOR360638-250MG
3-Amino-5-cyanophenylboronic acid 98%
3-Amino-5-cyanophenylboronic acid
Formula:
C₇H₇BN₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD03095195 CAS Number: 913943-05-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...