3-Carboxy-2-chlorophenylboronic acid 98%

Supplier: Apollo Scientific

OR360867-1G OR360867-250MG
APOSOR360867-1GEA 1190 EUR
APOSOR360867-1G APOSOR360867-250MG
3-Carboxy-2-chlorophenylboronic acid 98%
3-Carboxy-2-chlorophenylboronic acid
Formula: C₇H₆BClO₄
MW: 200,384 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18783132
CAS Number: 1314264-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR