3-Cyano-5-methylphenylboronic acid, pinacol ester 95+%
Supplier: Apollo Scientific
OR361035-1G
OR361035-250MG
APOSOR361035-1GEA
171
EUR
APOSOR361035-1G
APOSOR361035-250MG
3-Cyano-5-methylphenylboronic acid, pinacol ester 95+%
3-Cyano-5-methylphenylboronic acid, pinacol ester
Formula:
C₁₄H₁₈BNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD18383798 CAS Number: 1220219-59-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...