7-Bromoquinoline-3-carboxylic acid
Supplier: Apollo Scientific
|
Warning
|
OR3616-1G
OR3616-5G
APOSOR3616-1GEA
267
EUR
APOSOR3616-1G
APOSOR3616-5G
7-Bromoquinoline-3-carboxylic acid
7-Bromoquinoline-3-carboxylic acid
Formula:
C₁₀H₆BrNO₂ MW: 252,064 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08437554 CAS Number: 892874-34-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...