2-((tert-Butyldimethylsilanyl)ethynyl) boronic acid, pinacol ester

Supplier: Apollo Scientific

OR361614-1G OR361614-250MG
APOSOR361614-1GEA 360 EUR
APOSOR361614-1G APOSOR361614-250MG
2-((tert-Butyldimethylsilanyl)ethynyl) boronic acid, pinacol ester
2-((tert-Butyldimethylsilanyl)ethynyl) boronic acid, pinacol ester
Formula: C₁₄H₂₇BO₂Si
MW: 266,259 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09265092
CAS Number: 1073355-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR