3-Isopropylpyridin-2-amine ≥98%

Supplier: Apollo Scientific

OR361720-100MG OR361720-250MG
APOSOR361720-100MGEA 427 EUR
APOSOR361720-100MG APOSOR361720-250MG
3-Isopropylpyridin-2-amine ≥98%
3-Isopropylpyridin-2-amine
Formula: C₈H₁₂N₂
MW: 136.194 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19648929
CAS Number: 1101060-79-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR