1-Methyl-7-phenylpyrrolo[1,2-a]pyrimidin-2(1H)-one ≥98%
Supplier: Apollo Scientific
OR361723-100MG
OR361723-500MG
APOSOR361723-100MGEA
489
EUR
APOSOR361723-100MG
APOSOR361723-500MG
1-Methyl-7-phenylpyrrolo[1,2-a]pyrimidin-2(1H)-one ≥98%
1-Methyl-7-phenylpyrrolo[1,2-a]pyrimidin-2(1H)-one
Formula:
C₁₄H₁₂N₂O MW: 224,258 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD30187482 CAS Number: 61900-80-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...