(E)-4-(3-Chlorophenyl)-2-oxobut-3-enoic acid ≥98%
Supplier: Apollo Scientific
OR361724-100MG
OR361724-500MG
APOSOR361724-100MGEA
500
EUR
APOSOR361724-100MG
APOSOR361724-500MG
(E)-4-(3-Chlorophenyl)-2-oxobut-3-enoic acid ≥98%
(E)-4-(3-Chlorophenyl)-2-oxobut-3-enoic acid
Formula:
C₁₀H₇ClO₃ MW: 210,614 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19370468 CAS Number: 1244025-00-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...