4-Bromo-2-(methylthio)aniline ≥98%

Supplier: Apollo Scientific

OR361734-10G OR361734-1G
APOSOR361734-10GEA 1660 EUR
APOSOR361734-10G APOSOR361734-1G
4-Bromo-2-(methylthio)aniline ≥98%
4-Bromo-2-(methylthio)aniline
Formula: C₇H₈BrNS
MW: 218,114 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21194023
CAS Number: 475089-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR