5-Chloro-2-vinylbenzoic acid ≥98%

Supplier: Apollo Scientific

OR361735-100MG OR361735-1G OR361735-250MG
APOSOR361735-100MGEA 491 EUR
APOSOR361735-100MG APOSOR361735-1G APOSOR361735-250MG
5-Chloro-2-vinylbenzoic acid ≥98%
5-Chloro-2-vinylbenzoic acid
Formula: C₉H₇ClO₂
MW: 182.604 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30171263
CAS Number: 1467773-21-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR