N-Cyclopropyl-N-(2,3-dichlorobenzyl)acetamide ≥98%
Supplier: Apollo Scientific
OR361747-1G
OR361747-250MG
APOSOR361747-1GEA
691
EUR
APOSOR361747-1G
APOSOR361747-250MG
N-Cyclopropyl-N-(2,3-dichlorobenzyl)acetamide ≥98%
N-Cyclopropyl-N-(2,3-dichlorobenzyl)acetamide
Formula:
C₁₂H₁₃Cl₂NO MW: 258,144 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12828193 CAS Number: 1041439-18-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...