6-Chloro-4-methoxy indole
Supplier: Apollo Scientific
Synonyms:
6-Chloro-4-methoxy-1H-indole
OR3685T-100MG
OR3685T-250MG
OR3685T-1G
APOSOR3685T-100MGEA
205
EUR
APOSOR3685T-100MG
APOSOR3685T-250MG
APOSOR3685T-1G
6-Chloro-4-methoxy indole
6-Chloro-4-methoxy indole
A promutagen derived from fava beans.
Formula:
C₉H₈ClNO MW: 181,619 g/mol Melting Pt: 45 °C Storage Temperature: Ambient |
MDL Number:
MFCD00238560 CAS Number: 117970-23-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...