3-Amino-5-chloro-1-methyl-2(1H)-pyridinone ≥98%
Supplier: Apollo Scientific
Synonyms:
3-Amino-5-chloro-1-methylpyridin-2(1H)-one
OR370009-1G
OR370009-5G
APOSOR370009-1GEA
104
EUR
APOSOR370009-1G
APOSOR370009-5G
3-Amino-5-chloro-1-methyl-2(1H)-pyridinone ≥98%
3-Amino-5-chloro-1-methyl-2(1H)-pyridinone
Formula:
C₆H₇ClN₂O MW: 158,586 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28130270 CAS Number: 1441769-24-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...