4-Vinyl-dihydrofuran-2(3H)-one ≥98%

Supplier: Apollo Scientific

Synonyms: 4-vinyl-dihydrofuran-2(3H)-one

OR370031-100MG OR370031-250MG
APOSOR370031-100MGEA 420 EUR
APOSOR370031-100MG APOSOR370031-250MG
4-Vinyl-dihydrofuran-2(3H)-one ≥98%
4-Vinyl-dihydrofuran-2(3H)-one
Formula: C₆H₈O₂
MW: 112,127 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13181922
CAS Number: 53627-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR