2-(1-Benzylpyrrolidin-3-yl)pyridine ≥98%
Supplier: Apollo Scientific
OR370033-100MG
OR370033-1G
APOSOR370033-100MGEA
353
EUR
APOSOR370033-100MG
APOSOR370033-1G
2-(1-Benzylpyrrolidin-3-yl)pyridine ≥98%
2-(1-Benzylpyrrolidin-3-yl)pyridine
Formula:
C₁₆H₁₈N₂ Storage Temperature: Ambient |
MDL Number:
MFCD11042353 CAS Number: 145105-04-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...