5-Methyl-8-hydroxycoumarin ≥98%
Supplier: Apollo Scientific
OR370038-1G
OR370038-250MG
OR370038-500MG
APOSOR370038-1GEA
882
EUR
APOSOR370038-1G
APOSOR370038-250MG
APOSOR370038-500MG
5-Methyl-8-hydroxycoumarin ≥98%
5-Methyl-8-hydroxycoumarin
Formula:
C₁₀H₈O₃ MW: 176,169 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18450566 CAS Number: 36651-81-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...