8-Chloro-3,4-dihydronaphthalen-2(1H)-one ≥98%
Supplier: Apollo Scientific
OR370061-100MG
OR370061-250MG
APOSOR370061-100MGEA
148
EUR
APOSOR370061-100MG
APOSOR370061-250MG
8-Chloro-3,4-dihydronaphthalen-2(1H)-one ≥98%
8-Chloro-3,4-dihydronaphthalen-2(1H)-one
Formula:
C₁₀H₉ClO MW: 180,631 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04038576 CAS Number: 82302-27-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...