4-Iodo-3,5-dimethylbenzamide ≥98%

Supplier: Apollo Scientific

OR370081-1G OR370081-5G
APOSOR370081-1GEA 627 EUR
APOSOR370081-1G APOSOR370081-5G
4-Iodo-3,5-dimethylbenzamide ≥98%
4-Iodo-3,5-dimethylbenzamide
Formula: C₉H₁₀INO
MW: 275,086 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13180441
CAS Number: 1206679-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR