1,3-Benzoxazol-6-ol

Supplier: Apollo Scientific

OR370098-100MG OR370098-1G OR370098-250MG
APOSOR370098-100MGEA 104 EUR
APOSOR370098-100MG APOSOR370098-1G APOSOR370098-250MG
1,3-Benzoxazol-6-ol
1,3-Benzoxazol-6-ol
Formula: C₇H₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD18450414
CAS Number: 106050-81-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR