4-[Methoxy(methyl)carbamoyl]benzeneboronic acid 97%
Supplier: Apollo Scientific
Synonyms:
4-(N,O-Dimethylhydroxylaminocarbonyl)phenylboronic acid, 4-Borono-(N-methoxy-N-methyl)benzamide
OR3967-100G
OR3967-25G
OR3967-5G
APOSOR3967-100GEA
839
EUR
APOSOR3967-100G
APOSOR3967-25G
APOSOR3967-5G
4-[Methoxy(methyl)carbamoyl]benzeneboronic acid 97%
4-[Methoxy(methyl)carbamoyl]benzeneboronic acid
Formula:
C₉H₁₂BNO₄ MW: 209,007 g/mol Melting Pt: 118…124 °C Storage Temperature: Ambient |
MDL Number:
MFCD04115671 CAS Number: 179055-26-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...