2-Bromo-3-chlorophenol

Supplier: Apollo Scientific
Warning

OR399001-10G OR399001-1G OR399001-5G
APOSOR399001-10GEA 344 EUR
APOSOR399001-10G APOSOR399001-1G APOSOR399001-5G
2-Bromo-3-chlorophenol
2-Bromo-3-chlorophenol
Formula: C₆H₄BrClO
MW: 207,452 g/mol
Boiling Pt: 225 °C
Melting Pt: 55,5 °C
Storage Temperature: Ambient
MDL Number: MFCD04037216
CAS Number: 855836-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR