2-Bromo-4-nitrophenylacetonitrile

Supplier: Apollo Scientific

OR399027-1G OR399027-250MG
APOSOR399027-1GEA 393 EUR
APOSOR399027-1G APOSOR399027-250MG
2-Bromo-4-nitrophenylacetonitrile
2-Bromo-4-nitrophenylacetonitrile
Formula: C₈H₅BrN₂O₂
MW: 241,042 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12152243
CAS Number: 543683-48-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR