3-Bromo-5-nitrophenylacetonitrile 95%

Supplier: Apollo Scientific
Warning

OR400040-1G OR400040-250MG
APOSOR400040-1GEA 473 EUR
APOSOR400040-1G APOSOR400040-250MG
3-Bromo-5-nitrophenylacetonitrile 95%
3-Bromo-5-nitrophenylacetonitrile
Formula: C₈H₅BrN₂O₂
MW: 241,042 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18392325
CAS Number: 875819-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR