2-Bromo-3-methoxybenzoic acid
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Bromo-3-carboxyanisole, 2-Bromo-m-anisic acid
OR400074-1G
OR400074-5G
APOSOR400074-1GEA
49.5
EUR
APOSOR400074-1G
APOSOR400074-5G
2-Bromo-3-methoxybenzoic acid
2-Bromo-3-methoxybenzoic acid
Formula:
C₈H₇BrO₃ MW: 231,043 g/mol Melting Pt: 153…158 °C Storage Temperature: Ambient |
MDL Number:
MFCD11848447 CAS Number: 88377-29-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...