6-Hydroxyquinazolin-4(1H)-one

Supplier: Apollo Scientific

Synonyms: 6-Hydroxyquinazolin-4-ol, 6-Hydroxy-4-quinazolone

OR400075-1G OR400075-25G OR400075-5G
APOSOR400075-1GEA 78.3 EUR
APOSOR400075-1G APOSOR400075-25G APOSOR400075-5G
6-Hydroxyquinazolin-4(1H)-one
6-Hydroxyquinazolin-4(1H)-one
Formula: C₈H₆N₂O₂
MW: 162,145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05662723
CAS Number: 16064-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR