3-Bromo-2-iodophenol

Supplier: Apollo Scientific
Warning

OR400091-1G OR400091-250MG
APOSOR400091-1GEA 58 EUR
APOSOR400091-1G APOSOR400091-250MG
3-Bromo-2-iodophenol
3-Bromo-2-iodophenol
Formula: C₆H₄BrIO
MW: 298,904 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08166320
CAS Number: 855836-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR