2-Chloro-5-iodobenzyl bromide
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
2-(Bromomethyl)-1-chloro-4-iodobenzene
OR400200-1G
OR400200-5G
OR400200-25G
APOSOR400200-1GEA
26.1
EUR
APOSOR400200-1G
APOSOR400200-5G
APOSOR400200-25G
2-Chloro-5-iodobenzyl bromide
2-Chloro-5-iodobenzyl bromide
Formula:
C₇H₅BrClI MW: 331.376 g/mol Melting Pt: 94…95 °C Storage Temperature: Ambient |
MDL Number:
MFCD08236727 CAS Number: 793695-85-9 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...