2-Amino-4-iodophenol 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Hydroxy-5-iodoaniline
OR400203-1G
OR400203-250MG
OR400203-5G
APOSOR400203-1GEA
163
EUR
APOSOR400203-1G
APOSOR400203-250MG
APOSOR400203-5G
2-Amino-4-iodophenol 95%
2-Amino-4-iodophenol
Formula:
C₆H₆INO MW: 235,022 g/mol Melting Pt: 139 °C Storage Temperature: Ambient |
MDL Number:
MFCD14705890 CAS Number: 99969-17-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...