3-Bromo-5-methylphenylacetonitrile 95%

Supplier: Apollo Scientific
Warning

OR400218-1G OR400218-250MG
APOSOR400218-1GEA 655 EUR
APOSOR400218-1G APOSOR400218-250MG
3-Bromo-5-methylphenylacetonitrile 95%
3-Bromo-5-methylphenylacetonitrile
Formula: C₉H₈BrN
MW: 210,071 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18204796
CAS Number: 871116-91-5
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR