3-Bromo-6-chlorobenzene-1,2-diamine 95%

Supplier: Apollo Scientific
Warning

OR400228-1G OR400228-250MG
APOSOR400228-1GEA 17.6 EUR
APOSOR400228-1G APOSOR400228-250MG
3-Bromo-6-chlorobenzene-1,2-diamine 95%
3-Bromo-6-chlorobenzene-1,2-diamine
Formula: C₆H₆BrClN₂
MW: 221,482 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28040528
CAS Number: 1823876-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR