4-Chlorothiophene-2-carboxylic acid 95%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Carboxy-4-chlorothiophene
OR40031-250MG
OR40031-1G
OR40031-100MG
APOSOR40031-250MGEA
77
EUR
APOSOR40031-250MG
APOSOR40031-1G
APOSOR40031-100MG
4-Chlorothiophene-2-carboxylic acid 95%
4-Chlorothiophene-2-carboxylic acid
Formula:
C₅H₃ClO₂S MW: 162,594 g/mol Melting Pt: 138…140 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00082876 CAS Number: 59614-95-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...