3'-Bromo-5'-chloroacetophenone
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-(3-Bromo-5-chlorophenyl)ethanone
OR400317-1G
OR400317-5G
OR400317-25G
APOSOR400317-1GEA
84.4
EUR
APOSOR400317-1G
APOSOR400317-5G
APOSOR400317-25G
3'-Bromo-5'-chloroacetophenone
3'-Bromo-5'-chloroacetophenone
Formula:
C₈H₆BrClO MW: 233,49 g/mol Melting Pt: 29…32 °C Storage Temperature: Ambient |
MDL Number:
MFCD13189467 CAS Number: 154257-85-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...